Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAM1 Antibody (OTI3H5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245909
Description
Ensure accurate, reproducible results in Western Blot
VAM1 Monoclonal specifically detects VAM1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VAM1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
MAGUK protein p55T, membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6), p55T, protein associated with Lin7 2, VAM-1PALS2VAM1MAGUK p55 subfamily member 6, Veli-associated MAGUK 1 | |
Mouse | |
60.9 kDa | |
0.1 mL | |
Stem Cell Markers | |
51678 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI3H5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
MPP6 | |
Full length human recombinant protein of human MPP6(NP_057531) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction