Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582663 Supplier Invitrogen™ Supplier No. PA582663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEILEDITPL INVDENVAEL VGILKEPHFQ SLLEAHDIVA SKCYDSPPSS PEMNNSSINN QLLPVDAIRI LGIHKRAGEP LGVTFRVENN DLVIARIL Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.

Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like (see DLG1), ZO1-like (see TJP1), p55-like (see MPP1), and LIN2-like (see CASK), based on their size and the presence of additional domains. MPP6 is a member of the p55-like MAGUK subfamily (Tseng et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MPP6
Gene Accession No. Q9NZW5
Gene Alias CH474011:EDL88200; Dlgh4; Dlgh4 protein; Fibroblast growth factor-inducible protein 15; MAGUK p55 subfamily member 6; MAGUK protein p55T; membrane palmitoylated protein 6; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 6); membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); Mpp6; P55t; P55T protein; Pals2; protein associated with Lin7 2; protein associated with Lin-7 2; VAM1; VAM-1; VELI-associated MAGUK 1
Gene Symbols MPP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MPP6. Recombinant protein control fragment (Product #RP-92644).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51678
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.