Learn More
Invitrogen™ VAM1 Polyclonal Antibody

Description
Immunogen sequence: VKCHFDYNPY NDNLIPCKEA GLKFSKGEIL QIVNREDPNW WQASHVKEGG SAGLIPSQFL EEKRKAFVRR DWDNSGPFCG TISSKKKKKM MYLTTRNAEF DRHEIQIYEE VAKMPPFQRK TLVLIGAQGV GRRSLKNRFI VLNPTRFGTT VPFTSRKPRE DEKDGQAYKF VSRSEMEADI KAGKYLEHGE YEGNLYGTKI D; Positive Samples: 293T, SW480, K-562, Mouse brain, Mouse liver, Rat brain; Cellular Location: Membrane, Peripheral membrane protein.
Specifications
Specifications
| Antigen | VAM1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MPP6 |
| Gene Accession No. | Q9JLB0, Q9NZW5 |
| Gene Alias | CH474011:EDL88200; Dlgh4; Dlgh4 protein; Fibroblast growth factor-inducible protein 15; MAGUK p55 subfamily member 6; MAGUK protein p55T; membrane palmitoylated protein 6; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 6); membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); Mpp6; P55t; P55T protein; Pals2; protein associated with Lin7 2; protein associated with Lin-7 2; VAM1; VAM-1; VELI-associated MAGUK 1 |
| Gene Symbols | MPP6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.