Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen VAMP-2
Concentration 0.5 mg/mL
Dilution Western Blot 1.0-2.0 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224599SS
SDP
View Documents
Novus Biologicals
NBP224599SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224599
SDP
View Documents
Novus Biologicals
NBP224599
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

VAMP-2 Polyclonal specifically detects VAMP-2 in Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

VAMP-2
Western Blot 1.0-2.0 ug/ml
Polyclonal
Purified
RUO
Mouse, Rat
P63027
6844
A portion of amino acids 1-50 of rat VAMP-2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2)
VAMP2
IgG
Protein A purified
The SLAP amino acid sequence used as immunogen for the antibody is 100% homologous in human (isoforms a, b and c) and chimpanzee, 83% homologous in mouse and 78% homologous in rat and bovine.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.