Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP-2 Rabbit anti-Human, Clone: JM11-00, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP266774
Description
VAMP-2 Monoclonal specifically detects VAMP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
VAMP-2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation | |
FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2) | |
Rabbit | |
Protein A purified | |
Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
6844 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
JM11-00 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
VAMP2 | |
Synthetic peptide within Human VAMP-2 aa 9-42/ 116. (SwissProt: P63027 Human; SwissProt: P63044 Mouse; SwissProt: P63045 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction