Learn More
VAMP-5 Antibody (3E5), Janelia Fluor™ 549, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP250357JF549
Catalog No. NBP50357549
Only null left
Add to Cart
Description
Description
VAMP-5 Monoclonal specifically detects VAMP-5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
VAMP-5
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
VAMP5
GST-mVAMP-5-II and KLH-mVAMP-5-II peptide sequences
0.1 mL
Primary
Human
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
300000
Western Blot, Immunocytochemistry/Immunofluorescence
myobrevin, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin)
Mouse
Protein A purified
Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission
10791
Store at 4°C in the dark.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.