Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP-8 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232095G
Description
VAMP-8 Polyclonal antibody specifically detects VAMP-8 in Human samples. It is validated for Western BlotSpecifications
| VAMP-8 | |
| Polyclonal | |
| Western Blot | |
| EDBVAMP-8, Endobrevin, vesicle-associated membrane protein 8, vesicle-associated membrane protein 8 (endobrevin) | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Vesicle-Associated Membrane Protein 8 using the numbering given in entry NP_003752.2 (GeneID 8673). | |
| 0.1 mL | |
| Membrane Vesicle Markers | |
| 8673 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction