Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-8 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB266967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB266967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB266967 Supplier Novus Biologicals Supplier No. NBP232095IR
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAMP-8 Polyclonal antibody specifically detects VAMP-8 in Human samples. It is validated for Western Blot

Specifications

Antigen VAMP-8
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EDBVAMP-8, Endobrevin, vesicle-associated membrane protein 8, vesicle-associated membrane protein 8 (endobrevin)
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Vesicle-Associated Membrane Protein 8 using the numbering given in entry NP_003752.2 (GeneID 8673).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 8673
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.