Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP-8 Rabbit anti-Human, Mouse, Rat, Clone: 5H9S4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | VAMP-8 |
---|---|
Clone | 5H9S4 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
VAMP-8 Monoclonal antibody specifically detects VAMP-8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
VAMP-8 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EDBVAMP-8, Endobrevin, vesicle-associated membrane protein 8, vesicle-associated membrane protein 8 (endobrevin) | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VAMP-8 (Q9BV40). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5H9S4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Membrane Vesicle Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8673 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title