Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP1/VAMP2 Monoclonal Antibody (SP10)
GREENER_CHOICE

Catalog No. PIMA517608
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517608 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517608 Supplier Invitrogen™ Supplier No. MA517608
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17608 targets VAMP/synaptobrevin in ELISA, FACS, and WB applications and shows reactivity with Human samples. The MA5-17608 immunogen is human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96).

Vesicle-associated membrane protein 2 is essential for cAMP-regulated exocytosis. VAMP (also know as synaptobrevin) is part of the synaptic vesicle docking and fusion complex and plays a central role in neuroexocytosis. Two VAMP (vesicle-associated membrane protein) isoforms are expressed in the nervous system and are differently distributed among the specialized parts of the tissue. VAMP-1 and -2 are present in all tissues tested, including kidney, adrenal gland, liver, pancreas, thyroid, heart, and smooth muscle. The two isoforms are differentially expressed in various tissues and their level may depend on differentiation. VAMP-1 is restricted to exocrine pancreas and to kidney tubular cells, whereas VAMP-2 is the predominant isoform present in Langerhans islets and in glomerular cells. Both isoforms show a patchy vesicular intracellular distribution in confocal microscopy. Evidence for the importance of neuronal VAMPproteins in the physiology of all cells is indicated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP1/VAMP2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone SP10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene VAMP1
Gene Accession No. P23763, P63027
Gene Alias RATVAMPB; RATVAMPIR; SPAX1; SYB; SYB1; SYB2; synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin-1; Synaptobrevin-2; Vamp1; VAMP-1; VAMP2; VAMP-2; vesicle associated membrane protein 1; vesicle associated membrane protein 2; Vesicle-associated membrane protein (synaptobrevin 2); vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2)
Gene Symbols VAMP1, VAMP2
Host Species Mouse
Immunogen Human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96).
Purification Method Affinity chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6843, 6844
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.