Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP1 Monoclonal Antibody (SP10)
GREENER_CHOICE

Catalog No. PIMA547693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547693 Supplier Invitrogen™ Supplier No. MA547693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specific for endogenous levels of the ~16 kDa synaptobrevin. The antibody has also been demonstrated to work in immunohistochemistry on formalin fixed, vibratome sections, but does not work on paraffin sections.

Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone SP10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 10mM HEPES with 100 μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5
Gene Vamp1
Gene Accession No. P23763, Q0V7N0, Q62442
Gene Alias DKFZp686H12131; lew; MRXSYP; SPAX1; SYB1; Syb-1; synaptobrevin 1; synaptobrevin-1; vamp; Vamp1; VAMP-1; vesicle associated membrane protein 1; vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1)
Gene Symbols Vamp1
Host Species Mouse
Immunogen Synaptic immunoprecipitate (crude) from human brain.
Purification Method IgM
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22317, 513621, 6843
Target Species Human, Mouse, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.