Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597166 Supplier Invitrogen™ Supplier No. PA597166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSAPAQPPAE GTEGTAPGGG PPGPPPNMTS NRRLQQTQAQ VEEVVDIIRV NVDKVLERDQ KLSELDDRAD ALQAGASQFE SSAAKLKRKY WWKNCK; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell junction, Cytoplasmic vesicle, Cytoplasmic vesicle membrane, Mitochondrion outer membrane, Single-pass type IV membrane protein, secretory vesicle, synapse, synaptic vesicle membrane, synaptosome, synaptosome.

Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Vamp1
Gene Accession No. P23763, Q62442, Q63666
Gene Alias DKFZp686H12131; lew; MRXSYP; SPAX1; SYB1; Syb-1; synaptobrevin 1; synaptobrevin-1; vamp; Vamp1; VAMP-1; vesicle associated membrane protein 1; vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1)
Gene Symbols Vamp1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-96 of human VAMP1 (NP_9547401).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22317, 25624, 6843
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.