Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | VAMP1 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8500
![]()
|
Proteintech Group Inc
131151AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Synaptophysin is a 38kDa glycoprotein present in the membrane of neuronal presynaptic vesicles in brain, spinal cord, retina, vesicles of adrenal medulla, neuromuscular junctions, and endocrine cells. It is also expressed by neuroendocrine cells throughout the body, both normal and neoplastic. Synaptophysin is a useful marker for the identification of normal neuroendocrine cells and neuroendocrine neoplasms. This antibody reacts with neuroendocrine cells of human adrenal medulla, carotid body, skin, pituitary gland, thyroid, lung, pancreas, and gastrointestinal mucosa. It also reacts with a wide spectrum of neuroendocrine neoplasms of neural type including neuroblastomas, ganglioneuroblastomas, ganglioneuromas, pheochromocytomas, chromaffin, and non-chromaffin paragangliomas.Specifications
| VAMP1 | |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P23763, Q62442, Q63666 | |
| 22317, 25624, 6843 | |
| VAMP1 Fusion Protein Ag3787 | |
| Primary | |
| -20°C | |
| VAMP1 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| SYB1, Synaptobrevin 1, VAMP 1, VAMP1 | |
| VAMP1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title