Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP1/VAMP2/VAMP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103178 Supplier Invitrogen™ Supplier No. PA5103178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total VAMP 1/2/3.

Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP1/VAMP2/VAMP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VAMP1
Gene Accession No. P23763, P63024, P63025, P63027, P63044, P63045, Q15836, Q62442, Q63666
Gene Alias CEB; Cellubrevin; D130027G05Rik; lew; RATVAMPB; RATVAMPIR; SPAX1; SYB; SYB1; Syb-1; SYB2; Syb-2; SYB3; sybII; synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin II; synaptobrevin-1; Synaptobrevin-2; synaptobrevin-3; VAMP1; VAMP-1; VAMP2; VAMP-2; Vamp3; VAMP-3; vesicle associated membrane protein 1; vesicle associated membrane protein 2; vesicle associated membrane protein 3; vesicle-associated membrane 2; Vesicle-associated membrane protein (synaptobrevin 2); vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2); vesicle-associated membrane protein 3
Gene Symbols VAMP1, Vamp2, Vamp3
Host Species Rabbit
Immunogen A synthesized peptide derived from human VAMP 1/2/3, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22317, 22318, 22319, 24803, 25624, 29528, 6843, 6844, 9341
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.