Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585475
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585475 Supplier Invitrogen™ Supplier No. PA585475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (90%), Xenopus laevis (96%), Rhesus Monkey (100%), Bovine (99%). Positive Control: mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The vesicle associated membrane proteins (VAMP) or synaptobrevins are calcium binding proteins specific to eukaryotes. VAMPs, along with synaptosomal associated protein of 25 kDa (SNAP-25) and syntaxin, form the core complex of soluble NSF attachment protein receptor (SNARE) proteins that interact with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and alpha-SNAP. These membrane associated proteins play a key role in the regulation of vesicle membrane fusion with the plasma membrane. The Clostridium tetani neurotoxin is a metalloprotease with specificity for VAMP. In Alzheimer's disease, VAMP levels of all isoforms appear to be significantly lowered. It is suggested that VAMP-2 is a resident protein of the insulin-sensitive glucose transporter type 4 (GLUT4) compartment and that it is required for GLUT4 vesicle incorporation into the cell surface in response to insulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene VAMP2
Gene Accession No. P63027, P63044, P63045
Gene Alias RATVAMPB; RATVAMPIR; SYB; SYB2; Syb-2; sybII; Synaptobrevin; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin II; synaptobrevin-2; vamp; Vamp2; VAMP-2; vesicle associated membrane protein 2; vesicle-associated membrane 2; Vesicle-associated membrane protein (synaptobrevin 2); vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2)
Gene Symbols VAMP2
Host Species Rabbit
Immunogen Full length human VAMP2 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22318, 24803, 6844
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.