Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP3/Cellubrevin Antibody (4G9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP19794850UL
Description
VAMP3/Cellubrevin Monoclonal specifically detects VAMP3/Cellubrevin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation.Specifications
VAMP3/Cellubrevin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry, ELISA 1:100-1:2000 | |
CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human VAMP3 mAb, clone k2A2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human cellubrevin protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, Flow Cytometry, ELISA | |
4G9 | |
Unconjugated | |
NP_004772 | |
VAMP3 | |
Recombinant human cellubrevin (1-77aa) purified from E. coli | |
50 μL | |
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
9341 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction