Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP3/Cellubrevin Antibody (4G9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197948F
Description
VAMP3/Cellubrevin Monoclonal specifically detects VAMP3/Cellubrevin in Human, Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
VAMP3/Cellubrevin | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
VAMP3 | |
Recombinant human cellubrevin (1-77aa) purified from E. coli | |
100 μL | |
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
9341 | |
Human, Mouse | |
Purified |
Flow Cytometry, ELISA | |
4G9 | |
Flow Cytometry, ELISA | |
CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human VAMP3 mAb, clone k2A2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human cellubrevin protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction