Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP3/Cellubrevin Antibody - Azide and BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
$158.60 - $478.00
Specifications
Antigen | VAMP3/Cellubrevin |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017963
![]() |
Novus Biologicals
NB100989250.1MG |
0.1 mg |
Each for $158.60
|
|
|||||
NB10098925
![]() |
Novus Biologicals
NB10098925 |
0.5 mg |
Each for $478.00
|
|
|||||
Description
VAMP3/Cellubrevin Polyclonal specifically detects VAMP3/Cellubrevin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
VAMP3/Cellubrevin | |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot | |
Unconjugated | |
Sheep | |
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
Q15836 | |
9341 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Mouse, Rat, Human | |
CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
VAMP3 | |
Primary | |
VAMP3/Cellubrevin |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title