Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP3/Cellubrevin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB300510
Description
VAMP3/Cellubrevin Polyclonal specifically detects VAMP3/Cellubrevin in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
VAMP3/Cellubrevin | |
Polyclonal | |
Unconjugated | |
PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 | |
P63024 | |
VAMP3 | |
Sythetic peptide corresponding to residues M(1) S T G V P S G S S A A T G S N R R(18) C of mouse VAMP-3. | |
100 μg | |
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission | |
9341 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction