Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VAMP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116218
Description
VAMP3 Polyclonal Antibody for Western Blot, IHC (P)
Synaptobrevins/VAMPs, syntaxins, and the 25-kDa synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Because of its high homology to other known VAMPs, its broad tissue distribution, and its subcellular localization, the protein encoded by this gene was shown to be the human equivalent of the rodent cellubrevin. In platelets the protein resides on a compartment that is not mobilized to the plasma membrane on calcium or thrombin stimulation.
Specifications
| VAMP3 | |
| Polyclonal | |
| Unconjugated | |
| VAMP3 | |
| CEB; Cellubrevin; D130027G05Rik; hypothetical protein LOC415163; Syb3; Synaptobrevin-3; vamp; Vamp3; VAMP-3; vesicle associated membrane protein 3; vesicle-associated membrane protein 3; vesicle-associated membrane protein 3 (cellubrevin); zgc:86726 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 22319, 415163, 9341 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P63024, Q15836 | |
| VAMP3 | |
| KLH conjugated synthetic peptide between 15-43 amino acids from the Central region of human VAMP3. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Zebrafish | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction