Learn More
Invitrogen™ VAMP4 Polyclonal Antibody

Description
PA1-768 detects VAMP-4 from human and mouse samples. PA1-768 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~18 kDa protein representing VAMP-4 from PC3 cell extract. Immunofluorescent staining of VAMP-4 in CV-1 cells using PA1-768 results in a staining pattern consistent with VAMP-4 staining. Immunofluorescence staining with this antibody requires saponin permeabilization. The PA1-768 antigen is recombinant rat VAMP-4.
Specifications
Specifications
| Antigen | VAMP4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | VAMP4 |
| Gene Accession No. | O70480, O75379 |
| Gene Alias | D1Ertd147e; vamp; VAMP24; VAMP4; VAMP-4; vesicle associated membrane protein 4; vesicle-associated membrane protein 4 |
| Gene Symbols | VAMP4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.