Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP5 Monoclonal Antibody (3E5)
GREENER_CHOICE

Catalog No. PIMA527812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527812 Supplier Invitrogen™ Supplier No. MA527812
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: human VAMP5-transfected 293T Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of vesicles and cell membranes. The VAMP5 gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. This VAMP family member may participate in vesicle trafficking events that are associated with myogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative
Gene Vamp5
Gene Accession No. O95183
Gene Alias AF119384; Camp; HSPC191; Myobrevin; vamp; Vamp5; VAMP-5; vesicle associated membrane protein 5; vesicle-associated membrane protein 5
Gene Symbols Vamp5
Host Species Mouse
Immunogen Human VAMP5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10791
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.