Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP5 Monoclonal Antibody (OTI7F2), TrueMAB™, OriGene
SDP

Catalog No. 502544079
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4079 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4079 Supplier OriGene Supplier No. CF808605
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VAMP5 Monoclonal antibody specifically detects VAMP5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen VAMP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7F2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ICOSLG
Gene Accession No. O75144
Gene Alias AF119384; Camp; HSPC191; Myobrevin; vamp; Vamp5; VAMP-5; vesicle associated membrane protein 5; vesicle-associated membrane protein 5
Host Species Mouse
Immunogen Full length human recombinant protein of human ICOSLG produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23308
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.