Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557148 Supplier Invitrogen™ Supplier No. PA557148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGIELERCQ QQANEVTEIM RNNFGKVLER GVKLAELQQR SDQLLDMSST FNKTTQNLAQ KKCWENIRYR Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.

Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of vesicles and cell membranes. The VAMP5 gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family and the SNARE superfamily. This VAMP family member may participate in vesicle trafficking events that are associated with myogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Vamp5
Gene Accession No. O95183
Gene Alias AF119384; Camp; HSPC191; Myobrevin; vamp; Vamp5; VAMP-5; vesicle associated membrane protein 5; vesicle-associated membrane protein 5
Gene Symbols Vamp5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VAMP5. Recombinant protein control fragment (Product #RP-96193).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10791
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.