Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116892 Supplier Invitrogen™ Supplier No. PA5116892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse lung, Mouse brain, Mouse kidney, Rat lung.

This gene encodes a transmembrane protein that is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family. The encoded protein localizes to late endosomes and lysosomes and is involved in the fusion of transport vesicles to their target membranes. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Vamp7
Gene Accession No. P51809, P70280, Q9JHW5
Gene Alias Sybl1; synaptobrevin-like 1; synaptobrevin-like protein 1; tetanus neurotoxin-insensitive VAMP; tetanus-insensitive VAMP; TIVAMP; Ti-VAMP; vamp; Vamp7; VAMP-7; vesicle associated membrane protein 7; vesicle-associated membrane protein 7
Gene Symbols Vamp7
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20955, 6845, 85491
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.