Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP8/Endobrevin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981330
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1330 Supplier Bioss Supplier No. BS11395R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.

Specifications

Antigen VAMP8/Endobrevin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VAMP8
Gene Accession No. Q9BV40
Gene Alias AU041171; BOS_11631; Edb; Endobrevin; LOC100338966; Vamp8; VAMP-8; vesicle associated membrane protein 8; Vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin); vesicle-associated membrane protein 8-like; wu:fa18d07; zgc:56588
Gene Symbols VAMP8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VAMP8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8673
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.