Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAMP8 Monoclonal Antibody (1414CT354.12.23.93)
GREENER_CHOICE

Catalog No. PIMA537661
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537661 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537661 Supplier Invitrogen™ Supplier No. MA537661
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VAMP8 Monoclonal Antibody for Western Blot, IHC (P)

Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAMP8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1414CT354.12.23.93
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene VAMP8
Gene Accession No. Q9BV40
Gene Alias AU041171; Edb; endobrevin; hypothetical protein LOC507309; vamp; VAMP8; VAMP-8; vesicle associated membrane protein 8; vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin); wu:fa18d07; zgc:56588
Gene Symbols VAMP8
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 2~24 amino acids from the N-terminal region of human VAMP8.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.