Learn More
VAMP8 Mouse anti-Human, Clone: 1414CT354.12.23.93, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human VAMP8.
Supplier: Abnova Corporation MAB12361
Description
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. It is associated with the perinuclear vesicular structures of the early endocytic compartment. It has been found that VAMP8 interacts specifically with the soluble NSF-attachment protein (alpha-SNAP), most likely through an VAMP8-containing SNARE complex. [provided by RefSeq]
Specifications
| VAMP8 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (1:25) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| EDB | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 8673 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1414CT354.12.23.93 | |
| Mouse monoclonal antibody raised against synthetic peptide of human VAMP8. | |
| In PBS (0.09% sodium azide) | |
| VAMP8 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 2∼24 at N-terminus region of human VAMP8. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.