Learn More
Van de Graaff Accessories
SDPSCI_ED
Catalog No. p-2367753
Click to view available options
Product Type:
Electric Plume
Catalog No. S462074
Supplier: Science First N125
Only null left
Add to Cart
Add to Cart
Accessories for Electrostatic Demonstrations.
Electric Plume
  • Can be mounted to the dome of the Van de Graaff with its rubber vacuum cup
  • Strips repel each other and stand up when charged
Electric Whirl
  • Consists of a 9.2 in. dia. (23cm) metal wheel with radial spokes and discharge points
  • Supported on a cone bearing
  • Wheel spins rapidly as electrons escape from the points when held near an electrostatic generator
Specifications
For Use With (Application) Used to acquired negative charge from the dome of Van de Graaff generator during for electrostatic demonstrations
For Use With (Equipment) Van de Graaff generator
Includes Colored ribbons attached to a 6cm dia. Rubbered vacuum cup
Product Type Electric Plume
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.