Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson Gloves Van Goat Impact Lined Gloves

Catalog No. 17099955
Click to view available options
Size:
Large
X-Large

Van Goat Impact Lined Gloves have 3M Thinsulate™ C100 lining for superior warmth in cold conditions.

  • 3M Thinsulate™ C100 lining with an additional layer of microfleece
  • Cutshield™ premium P-aramid/steel/polyester liner
  • Dryhide™ Oil and water resistant full-grain goatskin leather
  • Drivers style with inset thumb
  • Impact protection heavy duty flame resistant rubber on back of hand, fingers and wrist
  • Stitched with Aramid Fiber thread
  • Snug-fitting shirred elastic wrist
  • Slip-on style cuff

Specifications

Certifications/Compliance ANSI Cut, ANSI Impact, ANSI Puncture
Certifications/Compliance (Europe) EN388
Chemotherapeutic Rated No
Chlorinated No
Construction Cut and Sewn
Description P-aramid Blend Glove
Coating Material Uncoated
Color White, Red
Exterior Finish Smooth
Hand Ambidextrous
Material Goatskin Leather
Product Type Cold-resistant Gloves
Size Large
For Use With (Application) Winter, Oil and Gas, LNG, Industrial, Mechanic, Construction
Glove Cuff Style Elasticated
Liner Material Thinsulate
Lining Lined
Series Van Goat
Length (English) 9.5 in.
Length (Metric) 30 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.