Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadate-Molybdate Reagent, for Phosphorus Analysis, Ricca Chemical
SDP

Catalog No. p-2917148
Click to view available options
Quantity:
500 mL
1 L

APHA for phosphorus testing.

  • For phosphorus testing.

Specifications

pH Range 0 to 2.5
Volume (Metric) 500 mL
Packaging Natural Poly Bottle
Quantity 500 mL
Sterility Non-sterile
Test Method APHA for Phosphorus
Type Vanadate-Molybdate Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.