Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vanadium

Catalog No. 26293550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
Catalog No. Quantity
26-293-550G 50 g
26-293-510G 10 g
2 options

Catalog No. 26-293-550G

Supplier: Sigma Aldrich 26293550G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Vanadium

Specifications

CAS 7440-62-2
Melting Point 1890°C
Density 6.11 g/mL (at 25°C)
Boiling Point 3380°C
Molecular Formula V
Linear Formula V
Quantity 50 g
MDL Number MFCD00011453
Molecular Weight (g/mol) 50.94
Percent Purity 99.5% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.