Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Vanadium(III) 2,4-pentanedionate, typically 97%

Manufacturer:  Alfa Aesar™ 2294022

Quantity 100g

 View more versions of this product

Catalog No. AA2294022


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13476-99-8
MFCD00000033
Vanadium(III) acetylacetonate
348.27
UN3285
Vanadium(III) 2,4-pentanedionate

Specifications

13476-99-8
348.27
UN3285
100g
Vanadium(III) 2,4-pentanedionate
Powder
MFCD00000033
Vanadium(III) acetylacetonate
Air Sensitive
97% (typically)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit