Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium(III) chloride, 97%

Catalog No. p-221535
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

Vanadium(III) chloride, 97%, Cl3V, CAS Number-7718-98-1, Vanadium trichloride, 100g, 96% min. (Argentometry), Black to Blue, 3.0000g/mL, 231-744-6, 13,289, 157.3, GHS Signal Word: Danger | CAS: 7718-98-1 | Cl3V | 157.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-98-1
Molecular Formula Cl3V
Molecular Weight (g/mol) 157.29
MDL Number MFCD00011454
InChI Key HQYCOEXWFMFWLR-UHFFFAOYSA-K
Synonym Vanadium trichloride
PubChem CID 62647
IUPAC Name trichlorovanadium
SMILES [Cl-].[Cl-].[Cl-].[V+3]

Specifications

Melting Point 250.0°C
Color Black to Blue
Physical Form Powder
Quantity 5 g
Assay Percent Range 96% min. (Argentometry)
Linear Formula VCl3
Fieser 13,289
Solubility Information Solubility in water: vigorous reaction. Other solubilities: soluble in abs.alcohol and ether
Formula Weight 157.3
Percent Purity 97%
Packaging Glass bottle
Specific Gravity 3
Density 3.0000g/mL
Chemical Name or Material Vanadium(III) chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 231-744-6
RTECSNumber YW2800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.