Learn More
Vanadium(III) chloride, 99% (metals basis), Thermo Scientific Chemicals
Description
Vanadium(III) chloride acts as an intermediate in organic synthesis and in the manufacture of organic light emitting diodes. It acts as a precursor to other vanadium(III) complexes. Further, it is used in the preparation of vanadium dichloride and organovanadium compounds. In addition to this, it serves as a catalyst in the production of ethylene propylene diene monomer rubber and polyethylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 10 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Soluble in tetrahydrofuran,alcohol,ether and hydrocarbons. |
Formula Weight | 157.3 |
Sensitivity | Light and moisture sensitive |
Density | 3 g/mL |
Chemical Name or Material | Vanadium(III) chloride |
Safety and Handling
Hazard Category | H302-H314-H335 |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 231-744-6 |
RTECSNumber | YW2800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.