Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium(III) oxide, 99.7% (metals basis)

Catalog No. AA4337736
Click to view available options
Quantity:
100 g
500 g

CAS: 1314-34-7 | O3V2 | 149.88 g/mol

Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-34-7
Molecular Formula O3V2
Molecular Weight (g/mol) 149.88
MDL Number MFCD00011455
InChI Key KFAFTZQGYMGWLU-UHFFFAOYSA-N
Synonym vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis
PubChem CID 518710
IUPAC Name oxo(oxovanadiooxy)vanadium
SMILES O=[V]O[V]=O

Specifications

Melting Point 1970°C
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.7% (metals basis)
Solubility Information Slightly soluble in water.
Formula Weight 149.88
Density 4.87
Chemical Name or Material Vanadium(III) oxide
Hazard Category H302-H311-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P321-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-230-9
RTECSNumber YW3050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.