Vanadium(IV) oxide, 99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 2295709

Quantity 10g

 View more versions of this product

Catalog No. AA2295709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12036-21-4
    Physical Form -100 Mesh Powder
    Assay Percent Range 99% (metals basis)
    Molecular Formula O2V
    Formula Weight 82.94
    MDL Number MFCD00049702
    Chemical Name or Material Vanadium(IV) oxide

    Specifications

    Physical Form -100 Mesh Powder
    Formula Weight 82.94
    Quantity 10g
    EINECS Number 234-841-1
    Density 4.339g/mL
    Assay Percent Range 99% (metals basis)
    Molecular Formula O2V
    MDL Number MFCD00049702
    CAS 12036-21-4
    Chemical Name or Material Vanadium(IV) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit