Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium(IV) oxide phthalocyanine

Catalog No. AA3958704
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3958704 2 g
AA3958702 0.5 g
AA3958709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3958704 Supplier Thermo Scientific Chemicals Supplier No. 03958704
Add to Cart

CAS: 13930-88-6 | C32H16N8OV | 579.476 g/mol

It is used as an organic light-emitting diode intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13930-88-6
Molecular Formula C32H16N8OV
Molecular Weight (g/mol) 579.476
MDL Number MFCD00041966
InChI Key YRZZLAGRKZIJJI-UHFFFAOYSA-N
Synonym oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 %
PubChem CID 2735160
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2]

Specifications

Color Blue to Purple
Physical Form Crystalline
Quantity 2 g
UN Number UN3285
Solubility Information Insoluble in water.
Formula Weight 579.45
Chemical Name or Material Vanadium(IV) oxide phthalocyanine
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber 237-700-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.