Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium Pentoxide, Powder, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18614642 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-642 50 kg Fiber Drum
18-614-644 25 g Amber Glass Bottle
18-614-639 125 g Amber Glass Bottle
18-614-641 500 g Amber Glass Bottle
18-614-643 2.5 kg Amber Glass Bottle
18-614-640 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-614-642 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. V101050KGBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 1314-62-1

Specifications

CAS Max % 1
Quantity 50 kg
Percent Purity 99.5%
Grade Reagent
Packaging Fiber Drum
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.