Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanadium(V) oxide, 99.6+%, ACROS Organics™
Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
Specifications
Assay Percent Range | 99.60% min |
Linear Formula | V2O5 |
Merck Index | 15, 10107 |
Solubility Information | Solubility in water: 8g/L. Other solubilities: soluble in acids and alkalies,insoluble in alcohol |
Formula Weight | 181.88 |
Percent Purity | 99.6+% |
Density | 3.3500g/mL |
Packaging | Glass bottle |
Solubility | Solubility in water: 8g/L. Other solubilities: soluble in acids and alkalies, insoluble in alcohol |
Specific Gravity | 3.35 |
Show More |
Safety and Handling
GHS H Statement
Suspected of damaging the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes damage to organs through prolonged or re
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Danger
EINECSNumber : 215-239-8
RTECSNumber : YW2450000
TSCA : TSCA