Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beckman Coulter Gentamicin Calibrator
Shop All Beckman Coulter Products
Click to view available options
Quantity:
15 mL
Description
- Our broad and growing portfolio of high-performing Six Sigma assays is compatible with AU Chemistry Analyzers - and it offers exceptional clinical quality
- Load it and leave it with liquid ready-to-use reagents
- Trust results with standardized test menus, assay protocols and reference ranges providing commutable patient results across AU analyzers
- Emit 2,000
Specifications
Specifications
| Detectable Analytes | Gentamicin |
| For Use With (Equipment) | Beckman Coulter Analyzer |
| Type | Calibrator |
| Disposable | Limited-use |
| Includes | 1 x 5 mL Bottle and 5 x 2 mL Bottles |
| Quantity | 15 mL |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction