Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vancomycin HydroChloride, Molecular Biology Grade, PROMO, Thermo Scientific Chemicals

Catalog No. J6279006PM
Click to view available options
Quantity:
1 g
5 g

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic that prevents cell wall synthesis
  • Originally obtained from the bacterial strain Streptomyces Orientalis
  • Vancomycin binds to D-alanyl-D-alanine motifs in cell wall precursors and leads to cell death
  • It is active against some penicillin-resistant bacteria

Application

  • Vancomycin hydrochloride is intended for use in the research of infections and for antibiotic resistance testing
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1404-93-9
Molecular Formula C66H76Cl3N9O24
Molecular Weight (g/mol) 1485.72 g/mol
MDL Number MFCD03613611,MFCD03613611
InChI Key LCTORFDMHNKUSG-UHFFFAOYNA-N
PubChem CID 124080918
IUPAC Name 48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.23,6.214,17.18,12.129,33.010,25.034,39]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
SMILES Cl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder
Color White
Quantity 5 g
Sensitivity Hygroscopic
Beilstein 3704657
Merck Index 149929
Solubility Information Soluble in water; Slightly soluble in methanol, ethanol and dimethylsulfoxide.
Grade Molecular Biology
Chemical Name or Material Vancomycin hydrochloride
RTECSNumber YW4380000
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.