Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vancomycin Hydrochloride, MP Biomedicals™

Catalog No. MP21955401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21955401 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21955401 Supplier MP Biomedicals, Inc Supplier No. 02195540.1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indicated for the treatment of serious infections caused by methicillin-resistant strains. Vancomycin Hydrochloride, MP Biomedicals is a glycopeptide antibiotic that blocks bacterial cell wall biosynthesis at the level of peptidoglycan biosynthesis. It is

Vancomycin, from Streptomyces orientalis, block bacterial cell wall biosynthesis by inhibiting incorporation of terminal D-alanyl-D-alanine moieties of the NAM/NAG-peptides. It also alters bacterial-cell-membrane permeability and RNA synthesis. It is widely studied in cancer patients.

Vancomycin is effective against Gram-positive bacteria. It is only indicated for the treatment of serious, life-threatening infections that are unresponsive to other less-toxic antibiotics. In particular, it is used to treat infections caused by susceptible strains of methicillin-resistant (β-lactam-resistant) staphylococci bacteria. It should not be used to treat methicillin-sensitive Staphylococcus aureus, because it is inferior to penicillins such as nafcillin.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Powder
CAS 123409-00-7
pH 2.5 to 4.5 (5% aq. soln.)
Source Streptomyces orientalis
Potency ca. 1000μg/mg
Color Pink
Quantity 100 mg
Molecular Formula C66H257Cl2N9O24
Solubility Information Soluble in water (>100 mg/mL). Moderately soluble in dilute methanol. Insoluble in acetone, alcohols, and ether.
InChI Key ARNOCQCQWLXLRG-UHFFFAOYSA-N
SMILES C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.CC(C1=CC(=C(C=C1)OC2CC(C(C(O2)CO)O)O)OC3=CC=CC=C3)NC(=O)C.C(=O)O.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl
IUPAC Name azane;N-[1-[4-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-phenoxyphenyl]ethyl]acetamide;formic acid;methane;pentadecahydrate;dihydrochloride
Molecular Weight (g/mol) 1632.721
PubChem CID 44134997
Formula Weight 1485.7
Grade Not Available
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.