Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Vandetanib
GREENER_CHOICE

Catalog No. 749710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-971-0 10 mg
74-975-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-971-0 Supplier Tocris Bioscience™ Supplier No. 7497/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent VEGFR-2 inhibitor, also inhibits EGFR and RET oncoproteins activity

Vandetanib is a potent VEGFR-2 inhibitor (IC50 = 0.04 μM), and inhibitor of EGFR (IC50 = 0.5 μM). Vandetanib potently inhibits (IC50 = 100 nM) RET oncoprotein activity in human thyroid cancer. Vandetanib displays anti-angiogenic properties in human nasopharyngeal carcinoma in mice model. Vandetanib is orally available and effective in vivo.

Specifications

Chemical Name or Material N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl)methoxy]-4-quinazolinamine
CAS 443913-73-3
Quantity 10 mg
Target VEGF Receptor Inhibitors
Molecular Formula C22H24BrFN4O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.