Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vang-like protein 1 Polyclonal Antibody, Huabio

Catalog No. 502544847
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4847 Supplier HUABIO RESEARCH INC Supplier No. 09055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vang-like protein 1 Polyclonal antibody specifically detects Vang-like protein 1 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen Vang-like protein 1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 25% glycerol and 0.05% sodium azide; pH 7.4
Gene VANGL1
Gene Accession No. Q8TAA9
Gene Alias KAI1 COOH-terminal interacting tetraspanin; KAI1 C-terminal interacting tetraspanin; Kitenin; Loop-tail protein 2; loop-tail protein 2 homolog; LPP2; STB2; Stbm; STBM2; strabismus; strabismus 2; Van Gogh-like protein 1; vang, van gogh-like 1; VANGL planar cell polarity protein 1; Vangl1; vang-like 1 (van gogh, Drosophila); vang-like protein 1
Host Species Rabbit
Immunogen Synthetic peptide within Human Vang-like protein 1 aa 114-171.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.