Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vang-like Protein 1/VANGL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP288565
Description
Vang-like Protein 1/VANGL1 Polyclonal specifically detects Vang-like Protein 1/VANGL1 in Human samples. It is validated for Western Blot.Specifications
Vang-like Protein 1/VANGL1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
KAI1 C-terminal interacting tetraspanin, Loop-tail protein 2 homolog, LPP2, MGC5338, STB2KITENIN, STBM2, Strabismus 2, Van Gogh-like protein 1, vang (van gogh, Drosophila)-like 1, vang, van gogh-like 1 (Drosophila), vang-like 1 (van gogh, Drosophila), vang-like protein 1 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human Vang-like Protein 1/VANGL1 | |
100 μL | |
Wnt Signaling Pathway | |
81839 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction