Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vang-like Protein 2/VANGL2 Rabbit anti-Human, Mouse, Rat, Clone: 5N5B3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $516.00
Specifications
| Antigen | Vang-like Protein 2/VANGL2 |
|---|---|
| Clone | 5N5B3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Vang-like Protein 2/VANGL2 Monoclonal antibody specifically detects Vang-like Protein 2/VANGL2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Vang-like Protein 2/VANGL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Neuroscience, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 57216 | |
| IgG | |
| Affinity purified |
| 5N5B3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| KIAA1215Loop-tail protein 1 homolog, loop-tail-associated protein, LPP1, LTAPVan Gogh-like protein 2, MGC119403, MGC119404, STB1Strabismus 1, STBM, STBM1, vang (van gogh, Drosophila)-like 2, vang, van gogh-like 2 (Drosophila), vang-like 2 (van gogh, Drosophila), vang-like protein 2 | |
| A phospho specific peptide corresponding to residues surrounding S79/S82/S84 of human Vang-like Protein 2/VANGL2 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title