Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VANGL1 Monoclonal Antibody (CL0241)
GREENER_CHOICE

Catalog No. PIMA524592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524592 Supplier Invitrogen™ Supplier No. MA524592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: DTESTYSGYS YYSSHSKKSH RQGERTRERH KSPRNKDGRG SEKSVTIQPP TGEPLLGNDS TRTEEVQDDN WGETTTAITG TSEHSISQED IARISKDMED SVGLDCKRY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Vang-like 1 protein is a novel human homologue of Drosophila Van Gogh/Strabismus. The Vang-like 1 gene consists of at least 7 exons, and encodes a protein with four transmembrane domains and the C-terminal Ser/Thr-X-Val motif. Studies shows that the expression of the protein is significantly up-regulated in gastric cancer cell lines MKN28, MKN74, pancreatic cancer cell lines BxPC-3, PSN-1 and Hs766T. The protein may serve as an effector mediating the intestinal trefoil factor (ITF) healing response of the intestinal mucosa. The protein also serves as a diagnostic marker for hepatocellular carcinoma as well as a potential molecular target for development of novel therapeutic drugs. The pathological role of the protein has been identified in neural tube defects (NTD). Human Vang-like 1 gene maps to chromosome 1p11-p13.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VANGL1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL0241
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VANGL1
Gene Accession No. Q8TAA9
Gene Alias KAI1 COOH-terminal interacting tetraspanin; KAI1 C-terminal interacting tetraspanin; Kitenin; Loop-tail protein 2; loop-tail protein 2 homolog; LPP2; STB2; Stbm; STBM2; strabismus; strabismus 2; Van Gogh-like protein 1; vang, van gogh-like 1; VANGL planar cell polarity protein 1; Vangl1; vang-like 1 (van gogh, Drosophila); vang-like protein 1
Gene Symbols VANGL1
Host Species Mouse
Immunogen Recombinant protein fragment of Human VANGL1 (Product #RP-91180)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.