Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VANGL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101954 Supplier Invitrogen™ Supplier No. PA5101954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total VANGL1.

Vang-like 1 protein is a novel human homologue of Drosophila Van Gogh/Strabismus. The Vang-like 1 gene consists of at least 7 exons, and encodes a protein with four transmembrane domains and the C-terminal Ser/Thr-X-Val motif. Studies shows that the expression of the protein is significantly up-regulated in gastric cancer cell lines MKN28, MKN74, pancreatic cancer cell lines BxPC-3, PSN-1 and Hs766T. The protein may serve as an effector mediating the intestinal trefoil factor (ITF) healing response of the intestinal mucosa. The protein also serves as a diagnostic marker for hepatocellular carcinoma as well as a potential molecular target for development of novel therapeutic drugs. The pathological role of the protein has been identified in neural tube defects (NTD). Human Vang-like 1 gene maps to chromosome 1p11-p13.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VANGL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VANGL1
Gene Accession No. Q80Z96, Q8TAA9
Gene Alias KAI1 COOH-terminal interacting tetraspanin; KAI1 C-terminal interacting tetraspanin; Kitenin; Loop-tail protein 2; loop-tail protein 2 homolog; LPP2; STB2; Stbm; STBM2; strabismus; strabismus 2; Van Gogh-like protein 1; vang, van gogh-like 1; VANGL planar cell polarity protein 1; Vangl1; vang-like 1 (van gogh, Drosophila); vang-like protein 1
Gene Symbols VANGL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human VANGL1(Accession Q8TAA9), corresponding to amino acid residues R352-A402.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229658, 81839
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.