Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VANGL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | VANGL1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9649
![]() ![]() |
Proteintech Group Inc
146961AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Vang-like 1 protein is a novel human homologue of Drosophila Van Gogh/Strabismus. The Vang-like 1 gene consists of at least 7 exons, and encodes a protein with four transmembrane domains and the C-terminal Ser/Thr-X-Val motif. Studies shows that the expression of the protein is significantly up-regulated in gastric cancer cell lines MKN28, MKN74, pancreatic cancer cell lines BxPC-3, PSN-1 and Hs766T. The protein may serve as an effector mediating the intestinal trefoil factor (ITF) healing response of the intestinal mucosa. The protein also serves as a diagnostic marker for hepatocellular carcinoma as well as a potential molecular target for development of novel therapeutic drugs. The pathological role of the protein has been identified in neural tube defects (NTD). Human Vang-like 1 gene maps to chromosome 1p11-p13.1.Specifications
VANGL1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80Z96, Q8TAA9 | |
229658, 690366, 81839 | |
VANGL1 Fusion Protein Ag6306 | |
Primary | |
-20°C | |
VANGL1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Loop tail protein 2 homolog, LPP2, STB2, STBM2, Strabismus 2, Van Gogh like protein 1, Vang like protein 1, VANGL1 | |
Vangl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title